Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) Isopropoxide 98.0+%, TCI America™
SDP

Catalog No. Y00091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Y00091G 1 g
Y0009200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Y00091G Supplier TCI America Supplier No. Y00091G

Chemical Identifiers

CAS 2172-12-5
Molecular Formula C9H24O3Y
Molecular Weight (g/mol) 269.19
MDL Number MFCD00015641
InChI Key NREVZTYRXVBFAQ-UHFFFAOYSA-N
Synonym yttrium isopropoxide
PubChem CID 85308566
IUPAC Name propan-2-ol;yttrium
SMILES CC(C)O.CC(C)O.CC(C)O.[Y]

Specifications

Melting Point 290°C
Color White
Quantity 1 g
UN Number 1325
Formula Weight 266.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Yttrium(III) Isopropoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.