Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number AA1118018
Click to view available options
Quantity:
50 g
250 g
1 kg
1314-36-9
O3Y2
225.81
MFCD00011473
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
[O--].[O--].[O--].[Y+3].[Y+3]
1314-36-9
O3Y2
225.81
MFCD00011473
RUDFQVOCFDJEEF-UHFFFAOYSA-N
diyttrium(3+) trioxidandiide
[O--].[O--].[O--].[Y+3].[Y+3]

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In phosphors for color TV tubes, in microwave filters, and as a stabilizer for high-temperature service materials
TRUSTED_SUSTAINABILITY
Melting Point 2410°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.9% (REO)
Merck Index 14,10107
Solubility Information Insoluble in water; Soluble in alcohol and dilute acids
Formula Weight 225.81
Percent Purity 99.9%
Grade Reacton™
Density 5.01 g/mL
Assay (REO)
Chemical Name or Material Yttrium(III) oxide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-233-5
RTECSNumber ZG3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit