Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide, REacton™, 99.999% (REO)

Catalog No. aa1118214
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1118214 25 g
AA1118222 100 g
AA11182A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1118214 Supplier Thermo Scientific Chemicals Supplier No. 01118214
Only null left
Add to Cart

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.999% (REO)
Solubility Information Insoluble in water. Soluble in alcohol dilute acids
Formula Weight 225.81
Odor Odorless
Density 5.01
Chemical Name or Material Yttrium(III) oxide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-233-5
RTECSNumber ZG3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.