Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yttrium(III) oxide, Aerosol Refractory Paint, 99.9% (REO), Thermo Scientific™

Catalog No. AA40392C
Change view
Click to view available options
Quantity:
13 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40392C 13 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40392C Supplier Thermo Scientific Chemicals Supplier No. 40392:C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Provides Easy Application of Uniform, Thin, Protective High Temperature Y2O3 Coatings. Substrates Can Be Ceramic

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coverage: 30-50 ft.2/can. Carrier: acetone, propane, butane, alcohol. Flammability: Extremely flammable
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Quantity 13 oz.
Assay Percent Range 99.9% (REO)
Odor Alcohol-like
Appearance White aerosol
Chemical Name or Material Yttrium(III) oxide, Maximum temperature use is 1900°C air, vacuum or inert
Hazard Category H220-H319-H336: Extremely flammable gas. Causes serious eye irritation. May cause drowsiness or dizziness.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.