Learn More
Description
- Used to clean reagent probes
Specifications
Specifications
| Type | Reagent Probe Wash K |
| For Use With (Application) | Clean Reagent Probe |
| For Use With (Equipment) | Used on Yumizen C1200 Clinical Chemistry Analyzer |
| Quantity | 3 Pk. 500 mL Bottles |
| Height (English) | 6 in. |
| Height (Metric) | 15.24 cm |
| Width (English) | 6 in. |
| Width (Metric) | 15.25 cm |
| Voltage | 0/1 V |
| Weight (English) | 50.72 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.