Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam (Z)-4-Hydroxytamoxifen, Estrogen Receptor Modulator, Solid, 25 mg

Catalog No. 502583337
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3337 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3337 Supplier Abcam Supplier No. AB14194325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent and selective estrogen receptor antagonist with higher potency in vitro than (Z)-tamoxifen.

  • Molecular Weight: 387.5 Da
  • Purity: >98%
  • Form: Solid
  • Activity: Potent and selective estrogen receptor antagonist (IC50 = 7 nM); active metabolite of (Z)-tamoxifen (ab120656) with higher in vitro potency
  • Biological Effect: Shows antiproliferative effect (IC50 = 27 μM) and induces p21 nuclear localization
  • Storage: Store at -20°C

Specifications

Target Molecule Estrogen Receptor
Shipping Condition Ambient
Buffer 100% 4-Hydroxytamoxifen
Content And Storage Store at -20°C.
Form Solid
Formulation 100% 4-Hydroxytamoxifen
Quantity 25 mg
For Use With (Application) Estrogen receptor modulation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.