Learn More
Abcam (Z)-4-Hydroxytamoxifen, Estrogen Receptor Modulator, Solid, 5 mg
Description
- Potent and selective estrogen receptor antagonist (IC50 = 7 nM).
- Active metabolite of (Z)-tamoxifen.
- Exhibits antiproliferative effect (IC50 = 27 μM).
- Induces p21 nuclear localization.
- Molecular weight: 387.5 Da.
- Purity: >98%.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Estrogen Receptor |
| Form | Solid |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Formulation | 100% 4-Hydroxytamoxifen |
| Quantity | 5 mg |
| For Use With (Application) | Estrogen receptor modulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.