Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Z)-4-Hydroxytamoxifen, Thermo Scientific™

Catalog No. AC469501000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469501000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469501000 Supplier Thermo Scientific Chemicals Supplier No. 469501000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material (Z)-4-Hydroxytamoxifen
CAS 68047-06-3
Quantity 100 mg
Melting Point 137°C to 143°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C26 H29 N O2
Physical Form Crystalline Powder
Color White to Almost-white
Purity ≥98.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.