Learn More
Medchemexpress LLC (Z)-aconitic acid | 585-84-2 | MFCD00063184 | 99.3% | 174.11 | C6H6O6 | 100 MG

Supplier: Medchemexpress LLC HYW016814R100MG
(Z)-Aconitic acid is an analytical reference standard used for research and analytical applications, providing a well-characterized cis-isomer of aconitic acid (CAS 585-84-2) for chromatographic and spectroscopic assays.
- High purity: 99.3% (catalog listing; COA reports assay 98.96% and HPLC 99.13%).
- Molecular formula: C6H6O6.
- Molecular weight: 174.11 g/mol.
- Appearance: solid, white to light yellow.
- Available sizes: 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg, and 10 mM 1 mL in DMSO.
- Storage: powder -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
- Packaging: brown glass bottle to protect integrity.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.