Learn More
CPC Scientific Z-Asp(OMe)-Glu(OMe)-Val-DL-Asp(OMe)-FMK 0.5MG
SDP
Supplier:  CPC Scientific CASP061A
Encompass
ONE-LETTER SEQUENCE: Z-D(OMe)-E(OMe)-V-DL-D(OMe)-FMKMOLECULAR FORMULA: C30H41F1N4O12MOLECULAR WEIGHT:668.67STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [210344-95-9], RESEARCH AREA: ApoptosisREFERENCES: J. Ma et al., Br. J. Pharmacol., 124, 756 (1998); W. Liu et al., NeuroReport 9, 2609 (1998); E. Weimann et al., Leukemia Res., 23, 751 (1999); A. Kondratyev and K. Gale, Mol. Brain Res., 75, 216 (2000)
Catalog No. 50-210-9027