Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Z-DEVD-FMK Caspase-3 Inhibitor
GREENER_CHOICESDP

Catalog No. 80091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-091 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 80-091 Supplier Tocris Bioscience™ Supplier No. 8009/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Caspase-3 Inhibitor

Z-DEVD-FMK Caspase-3 Inhibitor is a cell-permeable, irreversible inhibitor of caspase-3/CPP32. It inhibits tumor cell apoptosis. In vivo, it has a neuroprotective effect in rat hippocampus following seizures. It also significantly reduces post-traumatic apoptosis, and improved neurological recovery, before and after induced traumatic brain injury in rat.

Specifications

Purity 95%
CAS 634911-78-7
Content And Storage Store at -20°C
Quantity 1 mg
Target Entry Caspase-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.