Learn More
Enzo Life Sciences Z-FA-FMK (Ready-to-Use) (100µl). CAS: 197855-65-5
SDP
Supplier:  Enzo Life Sciences ALX260148R100
Encompass_Preferred
Cell permeable negative control for FMK-labelled caspase inhibitors. Irreversible inhibitor of cathepsins B, L, and S, and likely cathepsin H, as well as cruzain and papain. Has been shown to inhibit effector, but not initiator, caspases in vitro, and suppress some forms of apoptosis. Has been used in vivo. Alternative name: Z-Phe-Ala-FMK, FMK caspase inhibitors negative control. MW: 386.4. Purity: ≥95% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8357