Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Z-GLY-PRO-ARG-7-AMINO-4-TRIFLUOROMETHYLCOUMARIN

Catalog No. MP3AFC06110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3AFC06110 10 mg
MP3AFC06125 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP3AFC06110 Supplier MP Biomedicals™ Supplier No. 03AFC06110
Add to Cart
Add to Cart

Used to test function of Granzyme A by using an enzymatic in vitro cell free assay.

Applications: Z-Gly-Pro-Arg-7-Amino-4-Trifluoromethylcoumarin is used to test function of Granzyme A by using an enzymatic in vitro cell free assay. Granzyme A is one of the most abundant granzymes known to be involved in cell death processes, using the alternative programmed cell death pathway.
Product Description: Z-Gly-Pro-Arg-7-Amino-4-Trifluoromethylcoumarin is a fluorescent substrate for granzyme A. Granzyme A is one of the most abundant granzymes known to be involved in cell death processes, using the alternative programmed cell death pathway.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage Keep cool and dry. Desiccate AFC-, AMC-, or MNA061 in solid form at room temperature. Store DMSO/DMF solution at -20 °C. Material is stable for at least one year, if stored as recommended.
Formula Weight 673.639
For Use With (Application) Substrate to test function of Granzyme A
Molecular Formula C31H34F3N7O7
Product Type Proteins, Enzymes & Peptides
Quantity 10 mg
Synonym Z-Gly-Pro-Arg-AFC, Z-GPR-AFC
Form Solid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.