Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPB™ Z-Link™ Respirator Kit, PX5 PAPR, Multi-Gas Cartridge, Zytec™ FR Face Seal
SDP

Z-Link™ provides versatility by serving a wide spectrum of industries and applications from welding and grinding, to chemical handling.

Supplier:  RPB™ 160022

Catalog No. 17-000-133

Alert:

WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.


Description

Description

  • Loose fitted respirator, no fit testing
  • APF 1000+
  • Anti-fog and scratch resistant ANSI Z87+ (EN166) lenses
  • Tear-off lens system ensures you always have a clear field of view
  • Air flow is fully adjustable with the air deflector system to suit personal preference and prevent fogging
  • Air supply circulates throughout head top, moving over skin for cool down
  • Fixed jaw ensures secure cape attachment
  • All parts can be purchased individually for easy maintenance and servicing
  • ANSI Z89.1 certified head protection (EN397).
  • Visor hinges open for easy communication with team (outside of respiratory hazards)
  • Padded interior with adjustable ratchet system
  • Compatible with RPB™ Comms-Link™ in-helmet communication system
  • Vision-Link™ light option available
  • Multiple breathing tube covers available to increase the life of the breathing tube
  • Welding visors and cape options are available
  • Deconable head suspension system and disposable hood
Specifications

Specifications

ANSI S3.19, Z89.1 Type 1 Class C, Z87+
8 to 12 hours
Polypropylene
COMBINATION
Yes
PAPR System for Heavy Industry
Yes
HEPA
No
Z-Link™
Yes
14 x 10.5 x 11.25 in.
1000
Lithium-ion
25 in.
TC-21C
Yes
One Size
99.9999% efficiency down to 0.12 microns
Z-Link™ Respirator (16-010-31), Zytec™ FR Chin Seal (16-731), Breathing Tube (04-831), PX5™ PAPR and Gas Door (03-802-FR), Multi-Gas/HE Cartridge/Filter (03-894), Welding Visor with ADF Lens (16-670)
Polycarbonate
1 Ea.
14°F to 113°F (-10°C to 45°C), <90%RH
3.326 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.