Learn More
RPB™ Z-Link™ Respirator Helmet with Tychem Face Seal C40

RPB Z-Link Respirator includes: 16-810 Safety Lens, 16-712 Face Seal Tychem QC, 04-830 Breathing Tube, 03-501 Climate Control Device (NIOSH Approved)
Supplier: RPB™ 1601512
Alert:
WARNING This product can expose you to chemicals including chromium, which is known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Description
- Loose fitted respirator, no fit testing. Allows for facial hair and glasses without compromising safety.
- APF 1000+
- Choice of 5 anti-fog and scratch resistant ANSI Z87+ (EN166) lenses: Premium Clear, Green Tint (Shade 5), Yellow, Black & Mirror Tint.
- Tear-off lens system ensures you always have a clear field of view.
- Air flow is fully adjustable with the air deflector system to suit personal preference and prevent fogging.
- Air supply circulates throughout head top, moving over your skin to cool you down.
- Fixed jaw ensures secure cape attachment.
- All parts can be purchased individually for easy maintenance and servicing.
- ANSI Z89.1 certified head protection (EN397).
- Visor hinges open for easy communication with team (outside of respiratory hazards).
- Padded interior with adjustable ratchet system.
- Compatible with RPB Comms-Link™ in-helmet communication system.
- Vision-Link™ light option available.
- Tychem™or Zytec™ cape options available.
- Multiple breathing tube cover available to increase the life of the breathing tube.
- Radiant heat options available.
- Welding visors and cape options are available to transform your Z-link and extend the product’s capabilities.
- Deconable head suspension system and disposable hood.
Specifications
ANSI S3.19, Z89.1 Type 1 Class C, Z87+ | |
ISO9001, NIOSH, ANSI S3.19, Z89.1, Z87+, CSEI CSA Z94.3-15, CE 2797 EN14594:2005 3 B, EN12941:1998, +A2:2008 TH3 P R SL, EN352-3:2002, EN397:2003+A1:2009, EN166:2001 CLASS 1 F, BSI Certified Product AS/NZS 1716:2012 BMP # 714303, AS/NZS 1801:1997, BMP # 714306, AS/NZS 1337.1:2010, BMP # 714304 | |
EN14594:2005 3 B, EN12941:1998, EN352-3:2002, EN397:2003+A1:2009, EN166:2001 CLASS 1 F | |
RPB Z-Link Respirator, includes: 16-810 Safety Lens, 16-712 Face Seal Tychem QC, 04-830 Breathing Tube, 03-501 Climate Control Device (NIOSH Approved) | |
HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA | |
PAPR Bump Cap Assembly | |
Yes | |
16-810 Safety Lens, 16-712 Face Seal Tychem QC, 04-830 Breathing Tube, 03-501 Climate Control Device (NIOSH Approved) | |
Polycarbonate | |
11 in. | |
14 in. |
NIOSH: APF 1000+, CE/AS/NZS: APF 40- TH3 NPF 500 | |
CSA Z94.3-15 | |
NIOSH PAPR & SAR | |
Gray, Green | |
Nylon 66 | |
Standard | |
Fabrication, Welding, Healthcare, Life Sciences, Painting, Coating, Pharma, Metal Cutting, Grinding, Brazing, Fettling, Thermospray, Sanding, Painting, Concerete Cutting, Aggregate, Chemical, Foundry | |
Anti-fog, Anti-scratch | |
Yes | |
11 in. | |
4 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.