Learn More
Description
- The peptide substrate contains a highly fluorescent 7-amino-4-methyl coumarin (AMC) group that is efficiently quenched by the amide bond formed between its amino group and the carboxyl group of the Arg residue
- Purity 99%±1% by HPLC
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | Bioactivity |
| Product Line | R&D Systems |
| Quantity | 10 mg |
| Format | Supplied as a stock solution in dimethyl sulfoxide (DMSO) |
| Molecular Weight (g/mol) | PredictedMoleMass: 578 Da |
| Product Type | Z: N-carbobenzyloxy; 7-Amino-4-methylcoumarin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
