Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 9 Peptide
Description
Specifications
Specifications
| Target | Serine/Threonine Kinases |
| For Use With (Application) | FRET |
| Assay | Serine/Threonine Kinase Assay |
| Content And Storage | Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. |
| Assay Entry | Biochemical phospho-specific activity (no antibody) |
| Label or Dye | Methylcoumarin |
| Product Type | Kinase Assay Kit |
| Quantity | 800 x 20 μL Assays |
| Format | 384-well plate |
| Sample Type | glycogen synthase I (PRPASVPP(pS)P(pS)) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.