Learn More
Bachem Z-Phe-Arg-pNA · HCl
SDP
Supplier:  Bachem 4009170.0250
Encompass_Preferred
Z-Phe-Arg-pNA · HCl, 59188-54-4, C29H33N7O6 · HCl, Mr 612.09, 250mg. Z-FR-pNA, a sensitive chromogenic substrate for cathepsin L and papain. Z-FR-pNA is also cleaved by falcipain 2. CAS Number (net): 117761-01-0.
Catalog No. NC1444071
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.