Learn More
Bachem Z-Phe-His-Leu-OH
SDP
Supplier:  Bachem 4000599.1000
Encompass_Preferred
Z-Phe-His-Leu-OH, 28458-19-7, C29H35N5O6, Mr 549.63, 1g. Substrate for angiotensin I-converting enzyme. Cleavage of Z-FHL by ACE yields His-Leu (G-2310), which can be quantified by generating a fluorescent adduct with o-phthalaldehyde.
Catalog No. 50-260-449