Learn More
Abcam Z-Prolyl-prolinal (Z-Pro-Pro-CHO), Prolyl endopeptidase inhibitor, 25MG
Supplier:  Abcam AB14171925MG
Encompass_Preferred
MW 330.4 Da, Purity >97%. Z-Prolyl-prolinal (Z-Pro-Pro-CHO), Prolyl endopeptidase inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6473