Learn More
Bachem Z-Arg-Arg-AMC
SDP
Supplier:  Bachem 4004789.0250
Encompass_Preferred
Z-Arg-Arg-AMC Hydrochloride, 88937-61-5, C30H39N9O6, Mr 621.70, 250mg. Z-RR-AMC, an excellent, specific substrate for cathepsin B, an enzyme found in secretory vesicles. Cathepsin B has been identified as a b-secretase, as such it is involved in b-amyloid production. Z-RR-AMC is a convenient fluorogenic substrate for stem bromelain, as it is scarcely affected by fruit bromelain.
Catalog No. 50-194-7019
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.