Learn More
Description
Biological Activity: Cell-permeable, irreversible pan-caspase inhibitor. Inhibits caspase processing and apoptosis induction in tumor cells in vitro (IC50 = 0.0015 - 5.8mM). Active in vivo.
Sequence:VAD(Modifications: Val-1 = Z-Val, Asp-3 = (OMe)-fluoromethylketone)
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Color | White |
| Product Type | Inhibitor |
| Target Proteins | Z-VAD-FMK |
| CAS | 187389-52-2 |
| Molecular Formula | C22H30FN3O7 |
| Formula Weight | 467.49g/mol |
| Solubility | Soluble to 9.35mg/mL in DMSO |
| Content And Storage | Store at 4°C |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 1 mg |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
