Learn More
Abcam Z-VAD(OH)-FMK, General Caspase Inhibitor, Solid, 5 mg
Description
- Molecular Weight: 453.5 Da
- Purity: >94%
- Form: Solid
- CAS Number: 220644-02-0
- Function: Irreversible general caspase inhibitor
- Storage Temperature: Store at -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | Caspase |
| Quantity | 5 mg |
| For Use With (Application) | Caspase inhibition, Immunoprecipitation, Functional studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.