Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Z-VAD(OMe)-Fluromethylketone Inhibitor

Catalog No. MP3FK00901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP3FK00901 1 mg
MP3FK00903 3 mg
MP3FK00905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP3FK00901 Supplier MP Biomedicals™ Supplier No. 03FK00901
Add to Cart
Add to Cart

The Z-VAD(OMe)-FMK inhibitor synthesized using proprietary technology is cell permeable and irreversibly binds to activated Caspases to block apoptosis.

Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).

TRUSTED_SUSTAINABILITY

Specifications

Color White
Content And Storage 4°C
Quantity 1 mg
Product Type Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor
Molecular Weight (g/mol) 467.487
Form Solid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.