Learn More
Abcam Z-VAD(OMe)-FMK, Pan-Caspase Inhibitor, Solid, 1 mg
Description
- Molecular Weight: 467.5 Da
- Purity: >90%
- Form: Solid
- Function: Cell permeable, irreversible pan-caspase inhibitor
- Storage Temperature: Store at -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Content And Storage | Store at -20°C. |
| Target Molecule | Caspase |
| Shipping Condition | Ambient |
| Form | Solid |
| Formulation | 90% Z-VAD(OMe)-FMK |
| Quantity | 1 mg |
| For Use With (Application) | Caspase inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.