Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Z-VAD(OMe)-FMK, Pan-Caspase Inhibitor, Solid, 5 mg

Catalog No. 502584591
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4591 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4591 Supplier Abcam Supplier No. AB1204875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly validated and pure pan-caspase inhibitor for research applications.

  • Molecular Weight: 467.5 Da
  • Form: Solid
  • Purity: >90%
  • Description: Cell permeable, irreversible pan-caspase inhibitor
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice
  • Citation Count: 22

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Target Molecule Caspase
Quantity 5 mg
For Use With (Application) Caspase inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.