Learn More
CPC Scientific Z-Val-Glu-Ile-Asp-AFC 100MG
SDP
Supplier:  CPC Scientific CASP074D
Encompass
ONE-LETTER SEQUENCE: ZVEID-AFCMOLECULAR FORMULA: C38H44F3N5O12MOLECULAR WEIGHT:819.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [219138-06-4], RESEARCH AREA: ApoptosisREFERENCES: N.A. Thornberry, and Y. Lazebnik, Science 281, 1312 (1998); R.V. Talanian, et al., J. Biol. Chem. 272, 9677 (1997)
Catalog No. 50-210-9057
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.