Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals™

Catalog No. MP03FK01501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
3 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP03FK01501 1 mg
MP03FK01503 3 mg
MP03FK01505 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP03FK01501 Supplier MP Biomedicals™ Supplier No. 03FK01501

Irreversibly inhibits activity of VEID-dependent caspases (e.g., caspase-6). Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals is a synthetic peptide designed as a methyl ester to facilitate cell permeability. It is used for both in vitro and

If the intended use of this product is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Z-Val-Glu(OMe)-Ile-Asp(OMe)-FMK used for both in vitro and in vivo studies. If the intended use is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups.
Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 652
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.