Learn More
AdipoGen Z-VRPR-FMK
SDP
Supplier:  AdipoGen AGCP30008C500
Encompass_Preferred
Chemical. CAS 1381885-28-4 free base. Formula C31H49FN10O6 . C2HF3O2. MW 676.8 . 114.0. Synthetic. Selective cell permeable and irreversible MALT1 inhibitor. Inhibits NF-kappaB-dependent gene expression and affects the growth, proliferation and survival of activated B cell-like diffuse large B cell lymphomas ABC-DLBCL and germinal center B cell like diffuse large B cell lymphoma GCB-DLBCL cell lines. Shown to inhibit the autoprocessing activity of MCA2ac calcium dependent in Trypanosoma brucei.
Catalog No. 50-256-2457
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.