Learn More
Tocris Bioscience™ Z-VRPR-FMK trifluoroacetate salt
Description
This peptide is supplied in gross weight.
Specifications
Specifications
| Gene ID (Entrez) | 10892 |
| Content And Storage | Store at -20°C |
| Description | MALT1 |
| Molecular Weight (g/mol) | Observed M.W.: 790.81 |
| Quantity | 500 μg |
| Structural Form | C31H49FN10O6.CF3CO2H |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.