Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Z-YVAD-FMK, Irreversible Caspase-1 Inhibitor, Lyophilized, 5 mg

Catalog No. 502583361
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3361 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3361 Supplier Abcam Supplier No. AB1413885MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly pure and validated reversible caspase-1 inhibitor with 98% purity.

  • Function: Irreversible caspase-1 inhibitor
  • Purity: >98%
  • Molecular Weight: 630.7 Da
  • Form: Lyophilized
  • Citation Count: 12

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Caspase-1
Substrate Properties Chemical Substrate
Form Lyophilized
Alternate Part Number AB141388
Quantity 5 mg
For Use With (Application) Caspase-1 inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.