Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zap-70 Monoclonal Antibody (ZAP-03)
GREENER_CHOICE

Catalog No. 01676183
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-183 Supplier Invitrogen™ Supplier No. MA119443
  • $566.00
    $465.60 / Each of 1

    Save $100.40

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with ZAP70, a 70 kDa protein tyrosine kinase expressed in T and NK cells (intracellular antigen). ZAP70 is a molecule susceptible to degradation. It is recommended to use freshly prepared cell lysates (protease inhibitors are essential) to avoid non-specific staining of degradation products.

ZAP-70 is a cytosolic protein tyrosine kinase that is a member of the Syk family of proteins. ZAP-70 is expressed exclusively in T-cells and natural killer cells and is required for T-cell receptor activation. Upon T cell antigen receptor (TCR) engagement, ZAP70 is phosphorylated on tyrosines 292, 315 and 319 in the interdomain B, located between the SH2 and kinase domains. Phosphorylation of both tyrosines 315 (a Vav binding site) and 319 (a Lck binding site) enhances ZAP70 function in mediating lymphocyte signaling, while tyrosine 292 terminates the transient activation of ZAP70 and attentuates lymphocyte signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zap-70
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ZAP-03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene ZAP70
Gene Accession No. P43403, P43404
Gene Alias 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Selective T cell defect; Srk; STCD; STD; syk-related protein tyrosine kinase; syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; Tyrosine-protein kinase ZAP-70;TZK; TZK; ZA70; Zap70; ZAP-70; Zeta chain associated protein kinase 70kDa; zeta chain of T cell receptor associated protein kinase; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD
Gene Symbols ZAP70
Host Species Mouse
Immunogen Bacterially expressed fusion protein representing C-terminal part (160 amino acids) of human ZAP-70 with histidine tag.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22637, 7535
Target Species Human, Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.