Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 Mouse anti-Human, FITC, Clone: SBZAP, Southern Biotech™
SDP

Catalog No. p-7144833 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB10650-02S 25 μg
OB10650-02L 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OB10650-02S Supplier Southern Biotechnology Associates Supplier No. 1065002S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech cell signaling related antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen ZAP-70
Applications Flow Cytometry
Classification Monoclonal
Clone SBZAP
Concentration 0.1mg/mL
Conjugate FITC
Formulation Supplied in PBS/NaN3
Host Species Mouse
Immunogen Human ZAP-70 (a.a. 280-309)
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.