Learn More
Andwin Scientific ZAPCAP-CR NYLON 0.45
SDP
Supplier:  Andwin Scientific 74365
Encompass_Preferred
The ZapCap Disposable Bottle-Top Filters are ZapCap. This product is compatible to work with bottles with neck sizes of 33-45mm Sealing gasket.
Catalog No. NC9545319
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.