Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520455 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520455 Supplier Invitrogen™ Supplier No. PA520455
  • $625.00
    $494.65 / Each of 1

    Save $130.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Suggested positive controls are mouse small intestine or human small intestine tissue lysate. PA5-20455 can be used with blocking peptide PEP-0572. PA5-20455 detects ZBP1 in WB application with human samples.

Z-DNA binding protein 1 (ZBP1) belongs to a family of proteins that contain the Zalpha domain which binds specifically to left-handed Z-DNA and Z-RNA. ZBP1 was initially identified as a novel gene that was up-regulated in activated macrophages in mice bearing ascites tumors, suggesting that is may play a role in processes such as host response in neoplasia. More recent reports indicate that the cytosolic ZBP1 has can act like the toll-like receptor TLR9 by detecting cytosolic double-stranded (ds) DNA and trigger induction of type I interferon and other innate immune responses. It is thought that the binding of ZBP1 to dsDNA enhances its association with innate immune response proteins such as the IRF3 transcription factor and the serine/threonine kinase TBK1 (also known as NAK). Multiple isoforms of ZBP1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZBP1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ZBP1
Gene Accession No. Q8VDA5, Q9H171, Q9QY24
Gene Alias 2010010H03Rik; C20orf183; Dai; Dlm1; DLM-1; DNA-dependent activator of IFN-regulatory factors; DNA-dependent activator of IRFs; mZaDLM; RP4-718J7.1; Tumor stroma and activated macrophage protein DLM-1; Zbp1; Z-DNA binding protein 1; Z-DNA-binding protein 1
Gene Symbols ZBP1
Host Species Rabbit
Immunogen An 18 amino acid synthetic peptide from near the carboxy terminus of human ZBP1. The immunogen is located within the last 50 amino acids of ZBP1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171091, 58203, 81030
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.