Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116142 Supplier Invitrogen™ Supplier No. PA5116142
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZC3H11A.

Involved in nuclear mRNA export; probably mediated by association with the TREX complex. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H11A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ZC3H11A
Gene Accession No. O75152
Gene Alias 1110003F06Rik; 5730454B08Rik; G630041M05Rik; Kiaa0663; mKIAA0663; RGD1308290; ZC3H11A; ZC3HDC11A; Zc3hh11a; Zinc finger CCCH domain-containing protein 11A; zinc finger CCCH type containing 11A; zinc finger CCCH-type containing 11A; zinc finger CCCH-type domain containing 11A
Gene Symbols ZC3H11A
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZC3H11A(Accession O75152), corresponding to amino acid residues S768-S810.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9877
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.