Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZC3H11A Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7144221
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1850 10 μL
50-157-1849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1850 Supplier Bethyl Laboratories, Inc Supplier No. IHC00297T
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon and Crab-eating macaque

ZC3H11A (zinc finger CCCH domain-containing protein 11A), also known as KIAA0663 or ZC3HDC11A, is an 810 amino acid protein that contains 3 C3 H1-type zinc fingers. ZC3H11A is expressed in heart, brain, liver, skeletal muscle, kidney, pancreas, spleen, testis, ovary, fetal brain and fetal liver. The gene encoding ZC3H11A maps to human chromosome 1q32.1 and mouse chromosome 1 E4. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. The rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene on chromosome 1 which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1.

Specifications

Antigen ZC3H11A
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZC3H11A
Gene Accession No. O75152
Gene Alias Zinc finger CCCH domain-containing protein 11A, KIAA0663, ZC3HDC11A
Gene Symbols ZC3H11A
Host Species Rabbit
Immunogen Between 700 and 750
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9877
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.