Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3H12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522137 Supplier Invitrogen™ Supplier No. PA522137
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZC3H12A is an MCP1 -induced protein that acts as a transcriptional activator and causes cell death of cardiomyocytes, possibly via induction of genes associated with apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H12A
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ZC3H12A
Gene Accession No. Q5D1E8
Gene Alias BC036563; bifunctional endoribonuclease and deubiquitinase ZC3H12A; dJ423B22.1; Endoribonuclease ZC3H12A; MCP-1 treatment-induced protein; MCP-induced protein 1; MCPIP; MCPIP1; Monocyte chemotactic protein-induced protein 1; Reg1; Regnase-1; RGD:1306776}; RGD1306776; ribonuclease ZC3H12A; RP3-423B22.1; zc3h12a; zc3h12a {ECO:0000312; Zinc finger CCCH domain-containing protein 12A; zinc finger CCCH type containing 12A; zinc finger CCCH-type containing 12A
Gene Symbols ZC3H12A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZC3H12A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.