Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZC3HAV1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531650 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531650 Supplier Invitrogen™ Supplier No. PA531650
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The zinc finger antiviral protein (ZC3HAV1) is a CCCH type zinc finger protein that induces an innate immunity to infections by retrovirus by preventing the accumulation of viral RNAs in the cytoplasm and recruits the RNA processing exosome to degrade target RNAs, thereby inhibiting virus replication. ZC3HAV1 is localized in the cytoplasm at steady state, but shuttles between nucleus and cytoplasm in a XPO1-dependent manner. ZAP is a direct target gene of IRF3 action in cellular antiviral responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3HAV1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7
Gene Zc3hav1
Gene Accession No. Q7Z2W4
Gene Alias 1200014N16Rik; 2900058M19Rik; 9130009D18Rik; 9830115L13Rik; ADP-ribosyltransferase diphtheria toxin-like 13; ARTD13; CCCH-type zinc finger antiviral protein (Zap); D6Bwg1452e; FLB6421; Inactive Poly [ADP-ribose] polymerase 13; PARP13; PRO1677; rZAP; ZAP; ZC3H2; Zc3hav1; ZC3HDC2; zinc finger antiviral protein; Zinc finger CCCH domain-containing protein 2; zinc finger CCCH type, antiviral 1; zinc finger CCCH-type antiviral protein 1; zinc finger CCCH-type containing, antiviral 1; zinc finger CCCH-type, antiviral 1
Gene Symbols Zc3hav1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZC3HAV1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56829
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.