Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC12 Antibody, ProSci
SDP

Catalog No. 16904860
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-904-860 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-904-860 Supplier ProSci LLC Supplier No. 594050400
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZCCHC12 Polyclonal Antibody specifically detects PACRG in Human Samples. It is validated for Western Blot.

Specifications

Antigen ZCCHC12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution For WB starting dilution is: 1:1000
Formulation Supplied in PBS with 0.09% (W/V) sodium azide.
Gene Accession No. Q6PEW1
Gene Alias Zinc finger CCHC domain-containing protein 12, Smad-interacting zinc finger protein 1, ZCCHC12, SIZN1
Host Species Rabbit
Immunogen This ZCCHC12 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 374-402 amino acids from the C-terminal region of human ZCCHC12.
Purification Method This antibody is purified through a protein A column, followed by peptide affinity purification.
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170261
Target Species Human
Content And Storage Store at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Form Liquid
Isotype Ig
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.