Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZCCHC16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551880
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551880 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551880 Supplier Invitrogen™ Supplier No. PA551880
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDPATFHLLA QNLICNETNQ SGQFEKALAD PNQDEESVTD MMDNLPDLIT QCIQLDKKHS DRPELLQSET QLPLLASLIQ HQALFSPTDP PPKKGPIQLR EGQLPLTPAK RARQQETQLC LYCSQSGHFT RDCLAKRSRA PATTNNTAH Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 60%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC16
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RTL4
Gene Accession No. Q6ZR62
Gene Alias mammalian retrotransposon-derived protein 4; MAR3; Mar4; MART4; Retrotransposon Gag-like protein 4; RTL4; ZCCHC16; Zinc finger CCHC domain-containing protein 16; zinc finger CCHC-type containing 16; zinc finger, CCHC domain containing 16
Gene Symbols RTL4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZCCHC16. Recombinant protein control fragment (Product #RP-92269).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340595
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.