Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC17 Monoclonal Antibody (OTI7F9), TrueMAB™, OriGene
SDP

Catalog No. 501682877
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2877 Supplier OriGene Supplier No. TA809552
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZCCHC17 (zinc finger, CCHC domain containing 17), also known as PS1D (putative S1 RNA-binding domain protein), Pnn (Pinin)-interacting nucleolar protein or pNO40, is a 241 amino acid protein that associates with both Pinin and the 60S ribosomal subunit. Localizing to nucleolus, ZCCHC17 is ubiquitously expressed and has been suggested to play a role in ribosome maturation and biogenesis. ZCCHC17 contains one CCHC-type zinc finger, a S1 motif domain and exists as two alternatively spliced isoforms that map to human chromosome 1p35.2. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in Stickler syndrome, Parkinson′s disease, Gaucher disease and Usher syndrome.

Specifications

Antigen ZCCHC17
Applications Western Blot
Classification Monoclonal
Clone OTI7F9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZCCHC17
Gene Accession No. Q9NP64
Gene Alias HSPC251, PS1D, pNO40
Gene Symbols ZCCHC17
Host Species Mouse
Immunogen Full length human recombit protein of human ZCCHC17 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51538
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.