Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZCCHC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734063 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4063 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4063 Supplier Proteintech Group Inc Supplier No. 233741AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZCCHC8 may be involved in pre-mRNA splicing.

Specifications

Antigen ZCCHC8
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZCCHC8
Gene Accession No. Q6NZY4, Q9CYA6
Gene Alias ZCCHC8
Gene Symbols ZCCHC8
Host Species Rabbit
Immunogen ZCCHC8 Fusion Protein Ag19985
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55596, 70650
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.