Learn More
Medchemexpress LLC N-(4-methoxybenzoyl)-L-valyl-N-(trifluoro-oxopentan-3-yl)pyrrolidine-2-carboxamide | 171964-73-1 | 95.0% | 499.5 g/mol | C24H32F3N3O5 | 10 MG
Supplier: Medchemexpress LLC HY1925410MG
ZD-0892 is a small-molecule, selective inhibitor of neutrophil elastase with reported Ki values of 6.7 nM for human neutrophil elastase and 200 nM for porcine pancreatic elastase. It is supplied as a research-grade reagent (purity 95%) and is used in biochemical and enzymatic studies to probe elastase-dependent pathways.
- Selective inhibition of human neutrophil elastase (Ki ~6.7 nM).
- Also inhibits porcine pancreatic elastase (Ki ~200 nM).
- Suitable for biochemical and enzymatic assays.
- Provided at research-grade purity for reproducible results.
- Small-molecule compound with molecular weight approximately 499.5 g/mol.