Learn More
Selleck Chemical LLC ZD-4190
SDP
Supplier:  Selleck Chemical LLC S65432MG
Encompass_Preferred
ZD-4190 is a submicromolar inhibitor of VEGF RTK activity in vitro with IC50 values of 29 4 nM and 708 63 nM for KDR and Flt-1 respectively
Catalog No. 50-000-02660
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.