Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZD 7155 hydrochloride

Catalog No. 121150R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-115-0R 50 mg
12-111-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-115-0R Supplier Tocris Bioscience™ Supplier No. 1211/50
This item is not returnable. View return policy

Selective non-peptide AT1 antagonist

ZD 7155 hydrochloride is a potent and selective competitive antagonist for the angiotensin II type 1 (AT1) receptor. Displaces [125I]-angiotensin II binding with an IC50 value of 3.8 nM in guinea pig adrenal gland membranes. Orally active, and is more potent and longer-acting than the prototype AT1 antagonist, losartan.

Specifications

Chemical Name or Material 5,7-Diethyl-3,4-dihydro-1-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1,6-naphthyridin-2(1H)-one hydrochloride
CAS 146709-78-6
Assay Percent Range >98%
Quantity 50 mg
Solubility Soluble to 25mM in ethanol
Molecular Formula C26H26N6O.HCl
Formula Weight Observed MW: 474.99

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.