Learn More
Cayman Chemical ANTIBODY ML-417 10mg
SDP
Supplier:  Cayman Chemical 3658210 MG
Encompass_Preferred
A c-MET inhibitor (IC50s 0 4-2 nM for wild-type and mutant forms) selective for c-MET over a panel of 13 additional kinases (IC50s 22-7800 nM) but does inhibit Ron and FLT1 (IC50s 7 and 10 nM respectively) inhibits autophosphorylation of FGFR2 and PDGFRa in KATO III cells at 2 UM reduces proliferation in a panel of cancer cell lines including cells expressing constitutively active c-MET FGFR2 or PDGFR (IC50s 1 UM for all) reduces tumor growth in a GTL-16 gastric cancer mouse xenograft model (10-200 mg/kg)
Catalog No. 50-290-9570
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.