Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735370 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5370 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5370 Supplier Proteintech Group Inc Supplier No. 265451AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

26545-1-AP recognizes 48-55 kDa bands.

ZDHHC1 (Zinc Finger DHHC-Type Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include protein-cysteine S-palmitoyltransferase activity and palmitoyltransferase activity.

Specifications

Antigen ZDHHC1
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZDHHC1
Gene Accession No. Q8WTX9
Gene Alias C16orf1, DHHC 1, HSU90653, ZDHHC1, Zinc finger protein 377, ZNF377
Gene Symbols ZDHHC1
Host Species Rabbit
Immunogen ZDHHC1 Fusion Protein Ag24028
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29800
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.