Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC13, Mouse anti-Human, Clone: 26D1, Millipore Sigma™

Catalog No. p-7162986 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1498100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT1498100 Supplier MilliporeSigma Supplier No. MABT1498100UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ZDHHC13, clone 26D1, Cat. No. MABT1498, is a mouse monoclonal antibody that detects Palmitoyltransferase ZDHHC13 and has been tested for use in Western Blotting.

Palmitoyltransferase ZDHHC13 (UniProt: Q8IUH4; also known as EC: 2.3.1.22, Huntingtin-interacting protein 14-related protein, HIP14-related protein, Huntingtin-interacting protein HIP3RP, Putative MAPK-activating protein PM03, Putative NF-kappa-B-activating protein 209, Zinc finger DHHC domain-containing protein 13, DHHC-13) is encoded by the ZDHHC13 (also known as HIP14L, HIP3RP) gene (Gene ID: 54503) in human. DHHC-13 is a multi-pass membrane protein that serves as a palmitoyl acyltransferase (PAT) specific for a subset of neuronal proteins, including HD and GAD2 and may play a role in the pathogenesis of Huntington disease (HD). It is reported to be dysfunctional in the presence of the HD mutation and HIP-14 deficient mice are shown to develop features of HD. DHHC-13 also plays a role in cellular magnesium homeostasis. It contains a DHHC domain (aa 426-476 in isoform 1 and 250-300 in isoform 2), which is required for its palmitoyltransferase activity. (Ref.: Young, FB et al. (2011). Prog. Neurobiol. 97(2); 220-238).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC13
Applications Western Blot
Classification Monoclonal
Clone 26D1
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Palmitoyltransferase ZDHHC13;EC: 2.3.1.22;Huntingtin-interacting protein 14-related protein;HIP14-related protein;Huntingtin-interacting protein HIP3RP;Putative MAPK-activating protein PM03;Putative NF-kappa-B-activating protein 209;Zinc finger DHHC domain-containing protein 13;DHHC-13
Gene Symbols ZDHHC13;HIP14L;HIP3RP
Host Species Mouse
Immunogen MBP-tagged recombinant fragment corresponding to 111 amino acids from the N-terminal region of human Palmitoyltransferase ZDHHC13, isoform 2.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001001483
Test Specificity Clone 26D1 detects human Palmitoyltransferase ZDHHC13. It targets an epitope within 111 amino acids from the N-terminal region of isoforms 1 and 2. This sequence is conserved in both isoforms.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.