Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZDHHC15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101738 Supplier Invitrogen™ Supplier No. PA5101738
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZDHHC15.

The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC15
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZDHHC15
Gene Accession No. Q8BGJ0, Q96MV8
Gene Alias 6030457O13Rik; DHHC-15; membrane-associated DHHC15 zinc finger protein; MRX91; Palmitoyltransferase ZDHHC15; UNQ1969/PRO4501; Zdhhc15; Zinc finger DHHC domain-containing protein 15; zinc finger DHHC-type containing 15; zinc finger, DHHC domain containing 15; zinc finger, DHHC-type containing 15
Gene Symbols ZDHHC15
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZDHHC15(Accession Q96MV8), corresponding to amino acid residues H293-T337.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108672, 158866
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.